N-[2-(4-Chlorophenoxy)ethyl]-2-isopropylaniline

Supplier: Matrix Scientific

032021-500MG
101832-866EA 306.15 USD
101832-866
N-[2-(4-Chlorophenoxy)ethyl]-2-isopropylaniline
N-[2-(4-Chlorophenoxy)ethyl]-2-isopropylaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10688033
CAS Number: 1040681-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR