2-Isopropyl-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

032020-500MG
101832-864EA 306.15 USD
101832-864
2-Isopropyl-N-(3-methylbenzyl)aniline
2-Isopropyl-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁N
MDL Number: MFCD10688032
CAS Number: 1039821-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR