3-Isobutoxy-N-(3-isobutoxybenzyl)aniline

Supplier: Matrix Scientific

032016-500MG
101832-856EA 306.15 USD
101832-856
3-Isobutoxy-N-(3-isobutoxybenzyl)aniline
3-Isobutoxy-N-(3-isobutoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10688028
CAS Number: 1040681-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR