3-Isobutoxy-N-[2-(4-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032011-500MG
101832-846EA 306.15 USD
101832-846
3-Isobutoxy-N-[2-(4-methylphenoxy)ethyl]aniline
3-Isobutoxy-N-[2-(4-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10688023
CAS Number: 1040694-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR