4-Chloro-N-(cyclohexylmethyl)aniline

Supplier: Matrix Scientific

032009-500MG
101832-842EA 306.15 USD
101832-842
4-Chloro-N-(cyclohexylmethyl)aniline
4-Chloro-N-(cyclohexylmethyl)aniline

Hazard: Irritant/Stored under Argon

Formula: C₁₃H₁₈ClN
MDL Number: MFCD03211353
CAS Number: 127660-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR