2-Isobutoxy-N-[2-(phenethyloxy)benzyl]aniline

Supplier: Matrix Scientific

032007-500MG
101832-838EA 306.15 USD
101832-838
2-Isobutoxy-N-[2-(phenethyloxy)benzyl]aniline
2-Isobutoxy-N-[2-(phenethyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10688021
CAS Number: 1040694-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR