2-Isobutoxy-N-(4-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

032004-500MG
101832-832EA 461.54 USD
101832-832
2-Isobutoxy-N-(4-isopropoxybenzyl)aniline
2-Isobutoxy-N-(4-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10688018
CAS Number: 1040694-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR