N-(4-Fluorobenzyl)-2-isobutoxyaniline

Supplier: Matrix Scientific

032001-500MG
101832-826EA 306.15 USD
101832-826
N-(4-Fluorobenzyl)-2-isobutoxyaniline
N-(4-Fluorobenzyl)-2-isobutoxyaniline

Hazard: Irritant

Formula: C₁₇H₂₀FNO
MDL Number: MFCD10688015
CAS Number: 1040694-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR