N-[2-(4-Chloro-3-methylphenoxy)ethyl]-2-isobutoxyaniline

Supplier: Matrix Scientific

032000-500MG
101832-824EA 306.15 USD
101832-824
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-2-isobutoxyaniline
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-2-isobutoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO₂
MDL Number: MFCD10688014
CAS Number: 1040694-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR