N-(2-Methylphenethyl)-4-(phenethyloxy)aniline

Supplier: Matrix Scientific

031999-500MG
101832-822EA 306.15 USD
101832-822
N-(2-Methylphenethyl)-4-(phenethyloxy)aniline
N-(2-Methylphenethyl)-4-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO
MDL Number: MFCD10688013
CAS Number: 1040694-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR