N-[2-(3-Methylphenoxy)butyl]-4-(phenethyloxy)aniline

Supplier: Matrix Scientific

031995-500MG
101832-814EA 306.15 USD
101832-814
N-[2-(3-Methylphenoxy)butyl]-4-(phenethyloxy)aniline
N-[2-(3-Methylphenoxy)butyl]-4-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MW: 375.52 g/mol
MDL Number: MFCD10688009
CAS Number: 1040694-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR