N-[2-(1-Naphthyl)ethyl]-4-(phenethyloxy)aniline

Supplier: Matrix Scientific

031994-500MG
101832-812EA 306.15 USD
101832-812
N-[2-(1-Naphthyl)ethyl]-4-(phenethyloxy)aniline
N-[2-(1-Naphthyl)ethyl]-4-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₆H₂₅NO
MDL Number: MFCD10688008
CAS Number: 1040694-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR