N-(2,4-Dichlorobenzyl)-4-(phenethyloxy)aniline

Supplier: Matrix Scientific

031987-500MG
101832-798EA 306.15 USD
101832-798
N-(2,4-Dichlorobenzyl)-4-(phenethyloxy)aniline
N-(2,4-Dichlorobenzyl)-4-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₁H₁₉Cl₂NO
MDL Number: MFCD10688002
CAS Number: 1040693-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR