N-[2-(2,4-Dimethylphenoxy)ethyl]-4-(phenethyloxy)aniline

Supplier: Matrix Scientific

031986-500MG
101832-796EA 306.15 USD
101832-796
N-[2-(2,4-Dimethylphenoxy)ethyl]-4-(phenethyloxy)aniline
N-[2-(2,4-Dimethylphenoxy)ethyl]-4-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₄H₂₇NO₂
MW: 361.49 g/mol
MDL Number: MFCD10688001
CAS Number: 1040693-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR