N-(3,5-Dichlorobenzyl)-4-(2-methoxyethoxy)aniline
Supplier: Matrix Scientific
031984-500MG
101832-792EA
306.15
USD
101832-792
N-(3,5-Dichlorobenzyl)-4-(2-methoxyethoxy)aniline
N-(3,5-Dichlorobenzyl)-4-(2-methoxyethoxy)aniline
Hazard: Irritant
Formula:
C₁₆H₁₇Cl₂NO₂ |
MDL Number:
MFCD10687999 CAS Number: 1040693-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...