4-(2-Methoxyethoxy)-N-(4-propoxybenzyl)aniline

Supplier: Matrix Scientific

031981-500MG
101832-786EA 306.15 USD
101832-786
4-(2-Methoxyethoxy)-N-(4-propoxybenzyl)aniline
4-(2-Methoxyethoxy)-N-(4-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₃
MDL Number: MFCD10687996
CAS Number: 1040693-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR