4-Chloro-N-[2-(4-isopropylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031980-500MG
101832-784EA 306.15 USD
101832-784
4-Chloro-N-[2-(4-isopropylphenoxy)ethyl]aniline
4-Chloro-N-[2-(4-isopropylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10687995
CAS Number: 1040693-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR