4-Isopropoxy-N-(4-isopropylbenzyl)aniline

Supplier: Matrix Scientific

031976-500MG
101832-776EA 306.15 USD
101832-776
4-Isopropoxy-N-(4-isopropylbenzyl)aniline
4-Isopropoxy-N-(4-isopropylbenzyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO
MDL Number: MFCD10687991
CAS Number: 1039911-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR