N-(3-Isobutoxybenzyl)-4-isopropoxyaniline

Supplier: Matrix Scientific

031975-500MG
101832-774EA 306.15 USD
101832-774
N-(3-Isobutoxybenzyl)-4-isopropoxyaniline
N-(3-Isobutoxybenzyl)-4-isopropoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10687990
CAS Number: 1040692-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR