N-(2-Chlorophenyl)-N-propylamine

Supplier: Matrix Scientific

031973-500MG
101832-770EA 306.15 USD
101832-770
N-(2-Chlorophenyl)-N-propylamine
N-(2-Chlorophenyl)-N-propylamine

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD03210918
CAS Number: 55238-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR