4-Isopropoxy-N-(4-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

031972-500MG
101832-768EA 306.15 USD
101832-768
4-Isopropoxy-N-(4-isopropoxybenzyl)aniline
4-Isopropoxy-N-(4-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687988
CAS Number: 1040692-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR