4-Isopropoxy-N-(4-propoxybenzyl)aniline

Supplier: Matrix Scientific

031969-500MG
101832-762EA 306.15 USD
101832-762
4-Isopropoxy-N-(4-propoxybenzyl)aniline
4-Isopropoxy-N-(4-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687985
CAS Number: 1040692-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR