4-Isopropoxy-N-(4-methylbenzyl)aniline

Supplier: Matrix Scientific

031966-500MG
101832-756EA 306.15 USD
101832-756
4-Isopropoxy-N-(4-methylbenzyl)aniline
4-Isopropoxy-N-(4-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁NO
MDL Number: MFCD10687982
CAS Number: 1037146-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR