4-Isopropoxy-N-(2-phenoxyethyl)aniline

Supplier: Matrix Scientific

031965-500MG
101832-754EA 306.15 USD
101832-754
4-Isopropoxy-N-(2-phenoxyethyl)aniline
4-Isopropoxy-N-(2-phenoxyethyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁NO₂
MDL Number: MFCD10687981
CAS Number: 1040692-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR