N-[3-(2-Ethoxyethoxy)benzyl]-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031963-500MG
101832-750EA 306.15 USD
101832-750
N-[3-(2-Ethoxyethoxy)benzyl]-4-(2-phenoxyethoxy)aniline
N-[3-(2-Ethoxyethoxy)benzyl]-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₄
MW: 407.51 g/mol
MDL Number: MFCD10687979
CAS Number: 1040692-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR