N-[3-(Phenethyloxy)benzyl]-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031962-500MG
101832-748EA 306.15 USD
101832-748
N-[3-(Phenethyloxy)benzyl]-4-(2-phenoxyethoxy)aniline
N-[3-(Phenethyloxy)benzyl]-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₉H₂₉NO₃
MW: 439.56 g/mol
MDL Number: MFCD10687978
CAS Number: 1040692-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR