N-(2-Isopropoxybenzyl)-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031961-500MG
101832-746EA 306.15 USD
101832-746
N-(2-Isopropoxybenzyl)-4-(2-phenoxyethoxy)aniline
N-(2-Isopropoxybenzyl)-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₄H₂₇NO₃
MDL Number: MFCD10687977
CAS Number: 1040691-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR