2-Chloro-N-(3,5-dichlorobenzyl)aniline

Supplier: Matrix Scientific

031959-500MG
101832-742EA 306.15 USD
101832-742
2-Chloro-N-(3,5-dichlorobenzyl)aniline
2-Chloro-N-(3,5-dichlorobenzyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₀Cl₃N
MDL Number: MFCD10687975
CAS Number: 1040691-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR