4-(2-Phenoxyethoxy)-N-(2-propoxybenzyl)aniline

Supplier: Matrix Scientific

031957-500MG
101832-738EA 306.15 USD
101832-738
4-(2-Phenoxyethoxy)-N-(2-propoxybenzyl)aniline
4-(2-Phenoxyethoxy)-N-(2-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₄H₂₇NO₃
MDL Number: MFCD10687973
CAS Number: 1040691-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR