N-[4-(Hexyloxy)benzyl]-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031955-500MG
101832-734EA 306.15 USD
101832-734
N-[4-(Hexyloxy)benzyl]-4-(2-phenoxyethoxy)aniline
N-[4-(Hexyloxy)benzyl]-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₇H₃₃NO₃
MDL Number: MFCD10687971
CAS Number: 1040691-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR