N-(2-Ethoxybenzyl)-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031954-500MG
101832-732EA 306.15 USD
101832-732
N-(2-Ethoxybenzyl)-4-(2-phenoxyethoxy)aniline
N-(2-Ethoxybenzyl)-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₃
MDL Number: MFCD10687970
CAS Number: 1040691-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR