N-Ethyl-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031950-500MG
101832-724EA 306.15 USD
101832-724
N-Ethyl-4-(2-phenoxyethoxy)aniline
N-Ethyl-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₉NO₂
MDL Number: MFCD10687966
CAS Number: 1040691-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR