N-(4-Methylbenzyl)-4-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

031949-500MG
101832-722EA 306.15 USD
101832-722
N-(4-Methylbenzyl)-4-(2-phenoxyethoxy)aniline
N-(4-Methylbenzyl)-4-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₂H₂₃NO₂
MDL Number: MFCD10687965
CAS Number: 1040690-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR