N-(4-Isopropylbenzyl)-4-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

031946-500MG
101832-716EA 306.15 USD
101832-716
N-(4-Isopropylbenzyl)-4-(tetrahydro-2-furanylmethoxy)aniline
N-(4-Isopropylbenzyl)-4-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₇NO₂
MDL Number: MFCD10687962
CAS Number: 1040690-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR