N-[2-(4-Methylphenoxy)propyl]-4-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

031944-500MG
101832-712EA 306.15 USD
101832-712
N-[2-(4-Methylphenoxy)propyl]-4-(tetrahydro-2-furanylmethoxy)aniline
N-[2-(4-Methylphenoxy)propyl]-4-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₇NO₃
MDL Number: MFCD10687960
CAS Number: 1040690-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR