N-[2-(2,4-Dichlorophenoxy)ethyl]-4-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

031939-500MG
101832-702EA 306.15 USD
101832-702
N-[2-(2,4-Dichlorophenoxy)ethyl]-4-(tetrahydro-2-furanylmethoxy)aniline
N-[2-(2,4-Dichlorophenoxy)ethyl]-4-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₁₉H₂₁Cl₂NO₃
MDL Number: MFCD10687955
CAS Number: 1040690-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR