N-(Cyclohexylmethyl)-4-(heptyloxy)aniline

Supplier: Matrix Scientific

031932-500MG
101832-688EA 306.15 USD
101832-688
N-(Cyclohexylmethyl)-4-(heptyloxy)aniline
N-(Cyclohexylmethyl)-4-(heptyloxy)aniline

Hazard: Irritant

Formula: C₂₀H₃₃NO
MDL Number: MFCD10687948
CAS Number: 1040689-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR