N-(4-Fluorobenzyl)-4-(heptyloxy)aniline

Supplier: Matrix Scientific

031931-500MG
101832-686EA 306.15 USD
101832-686
N-(4-Fluorobenzyl)-4-(heptyloxy)aniline
N-(4-Fluorobenzyl)-4-(heptyloxy)aniline

Hazard: Irritant

Formula: C₂₀H₂₆FNO
MDL Number: MFCD10687947
CAS Number: 1040689-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR