4-(Isopentyloxy)-N-propylaniline

Supplier: Matrix Scientific

031930-500MG
101832-684EA 306.15 USD
101832-684
4-(Isopentyloxy)-N-propylaniline
4-(Isopentyloxy)-N-propylaniline

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687946
CAS Number: 1040689-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR