N-(4-Butoxybenzyl)-2-chloroaniline

Supplier: Matrix Scientific

031926-500MG
101832-676EA 306.15 USD
101832-676
N-(4-Butoxybenzyl)-2-chloroaniline
N-(4-Butoxybenzyl)-2-chloroaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10687942
CAS Number: 1040689-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR