4-(Isopentyloxy)-N-[4-(pentyloxy)benzyl]aniline

Supplier: Matrix Scientific

031925-500MG
101832-674EA 306.15 USD
101832-674
4-(Isopentyloxy)-N-[4-(pentyloxy)benzyl]aniline
4-(Isopentyloxy)-N-[4-(pentyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₃H₃₃NO₂
MDL Number: MFCD10687941
CAS Number: 1040689-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR