4-(Isopentyloxy)-N-(2-phenoxybutyl)aniline
Supplier: Matrix Scientific
031924-500MG
101832-672EA
306.15
USD
101832-672
4-(Isopentyloxy)-N-(2-phenoxybutyl)aniline
4-(Isopentyloxy)-N-(2-phenoxybutyl)aniline
Hazard: Irritant
Formula:
C₂₁H₂₉NO₂ |
MDL Number:
MFCD10687940 CAS Number: 1040689-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...