4-(Isopentyloxy)-N-[2-(4-methoxyphenoxy)propyl]aniline

Supplier: Matrix Scientific

031923-500MG
101832-670EA 306.15 USD
101832-670
4-(Isopentyloxy)-N-[2-(4-methoxyphenoxy)propyl]aniline
4-(Isopentyloxy)-N-[2-(4-methoxyphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₃
MW: 343.47 g/mol
MDL Number: MFCD10687939
CAS Number: 1040689-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR