N-(3-Chlorobenzyl)-4-(isopentyloxy)aniline

Supplier: Matrix Scientific

031922-500MG
101832-668EA 306.15 USD
101832-668
N-(3-Chlorobenzyl)-4-(isopentyloxy)aniline
N-(3-Chlorobenzyl)-4-(isopentyloxy)aniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD10687938
CAS Number: 1040689-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR