2-Chloro-N-[2-(4-methylphenoxy)butyl]aniline

Supplier: Matrix Scientific

031916-500MG
101832-656EA 306.15 USD
101832-656
2-Chloro-N-[2-(4-methylphenoxy)butyl]aniline
2-Chloro-N-[2-(4-methylphenoxy)butyl]aniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10687932
CAS Number: 1040688-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR