4-(Benzyloxy)-N-[3-(isopentyloxy)benzyl]aniline

Supplier: Matrix Scientific

031913-500MG
101832-650EA 306.15 USD
101832-650
4-(Benzyloxy)-N-[3-(isopentyloxy)benzyl]aniline
4-(Benzyloxy)-N-[3-(isopentyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10687929
CAS Number: 1040688-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR