4-(Benzyloxy)-N-[2-(2,5-dimethylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031910-500MG
101832-644EA 306.15 USD
101832-644
4-(Benzyloxy)-N-[2-(2,5-dimethylphenoxy)ethyl]aniline
4-(Benzyloxy)-N-[2-(2,5-dimethylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MW: 347.46 g/mol
MDL Number: MFCD10687926
CAS Number: 1040688-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR