4-(Benzyloxy)-N-(4-fluorobenzyl)aniline

Supplier: Matrix Scientific

031906-500MG
101832-636EA 306.15 USD
101832-636
4-(Benzyloxy)-N-(4-fluorobenzyl)aniline
4-(Benzyloxy)-N-(4-fluorobenzyl)aniline

Hazard: Irritant

Formula: C₂₀H₁₈FNO
MDL Number: MFCD10687922
CAS Number: 1040689-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR