N-[2-(2-Methylphenoxy)butyl]-4-propoxyaniline

Supplier: Matrix Scientific

031902-500MG
101832-628EA 306.15 USD
101832-628
N-[2-(2-Methylphenoxy)butyl]-4-propoxyaniline
N-[2-(2-Methylphenoxy)butyl]-4-propoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10687918
CAS Number: 1040688-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR