N-[2-(2,5-Dimethylphenoxy)ethyl]-4-propoxyaniline

Supplier: Matrix Scientific

031901-500MG
101832-626EA 306.15 USD
101832-626
N-[2-(2,5-Dimethylphenoxy)ethyl]-4-propoxyaniline
N-[2-(2,5-Dimethylphenoxy)ethyl]-4-propoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687917
CAS Number: 1040688-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR